Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IL-12 p40 (human) Recombinant, Thermo Scientific™

IL-12 p40 (Hu) Recombinant protein RP-75619 from Thermo Scientific™ specifically detects IL-12 p40 (Hu) in Not Applicable samples, and it is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 16.75
Name Human IL-12 p40 Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75617

 
pierce biotechnology inc
RP75617
10μg purified protein (0.18mg/mL) in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75618

 
pierce biotechnology inc
RP75618
100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $785.90

PIRP75619

 
pierce biotechnology inc
RP75619
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 16.75
Name Human IL-12 p40 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

IL-12 p40 (Hu) Recombinant protein RP-75619 from Thermo Scientific™ specifically detects IL-12 p40 (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75619 detects IL-12 p40 (human) in Not Applicable samples. RP-75619 has been successfully used in Control and Western Blot procedures. RP-75619 The immunogen is N/A.