Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IL-12 p35 (human) Recombinant, Thermo Scientific™

IL-12 p35 (Hu) Recombinant protein RP-75616 from Thermo Scientific™ specifically detects IL-12 p35 (Hu) in Not Applicable samples, and it is validated for Control and WB

$288.55 - $4,205.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-12 p35 Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75614

 
pierce biotechnology inc
RP75614
10μg purified protein in PBS with 50% glycerol and no preservative; pH 7.4 10μg Each for $288.55

PIRP75615

 
pierce biotechnology inc
RP75615
100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $797.50

PIRP75616

 
pierce biotechnology inc
RP75616
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $4,205.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-12 p35 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

IL-12 p35 (Hu) Recombinant protein RP-75616 from Thermo Scientific™ specifically detects IL-12 p35 (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75616 detects IL-12 p35 (human) in Not Applicable samples. RP-75616 has been successfully used in Control and Western Blot procedures. RP-75616 The immunogen is N/A.