Please login to your online account to display your discounted pricing

anti-IL-12 p35 (human) Recombinant, Thermo Scientific™


IL-12 p35 (Hu) Recombinant protein RP-75616 from Thermo Scientific™ specifically detects IL-12 p35 (Hu) in Not Applicable samples, and it is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75616

 View more versions of this product

 Disclaimers

in vitro research only

Catalog No. PIRP75616

  • / Each
 

Description & Specifications

Specifications

Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Host Species E. Coli
Conjugate Unlabeled
Immunogen N/A
Format Purified
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Name Human IL-12 p35 Recombinant Protein
Source E. coli
Cross Reactivity Human
Recombinant Yes
Quantity 1mg

IL-12 p35 (Hu) Recombinant protein RP-75616 from Thermo Scientific™ specifically detects IL-12 p35 (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Formulation: 1mg purified protein in PBS with 50% glycerol and no preservative
RP-75616 contains purified recombinant human IL-12 p35 protein with an N-terminal hexahistidine tag. RP-75616 may be used as a positive control in many immunological assays.