Please login to your online account to display your discounted pricing

anti-IL-1 Receptor Antagonist (human) Recombinant, Thermo Scientific™


IL-1 Receptor Antagonist (Hu) Recombinant protein RP-75580 detects IL-1 Receptor Antagonist (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75579

 View more versions of this product

 Disclaimers

in vitro research only

Catalog No. PIRP75579

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg purified protein in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0
For Use With (Application) Control, Western Blot
Host Species E. Coli
Conjugate Unlabeled
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Immunogen N/A
Recombinant Yes
Name Human IL-1 Receptor Antagonist Recombinant Protein
Cross Reactivity Human
Source E. coli
Quantity 100μg

IL-1 Receptor Antagonist (Hu) Recombinant protein RP-75580 detects IL-1 Receptor Antagonist (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 100μg purified protein in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0
RP-75579 contains purified recombinant human IL-1 Receptor Antagonist protein with an N-terminal hexahistidine tag. RP-75579 may be used as a positive control in many immunological assays.