Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IL-1 Receptor Antagonist (human) Recombinant, Thermo Scientific™

IL-1 Receptor Antagonist (Hu) Recombinant protein RP-75580 detects IL-1 Receptor Antagonist (Hu) in Not Applicable samples, and is validated for Control and WB

$797.50 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-1 Receptor Antagonist Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75580

 
pierce biotechnology inc
RP75580
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75579

 
pierce biotechnology inc
RP75579
100μg purified protein in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0 100μg Each for $797.50
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-1 Receptor Antagonist Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

IL-1 Receptor Antagonist (Hu) Recombinant protein RP-75580 detects IL-1 Receptor Antagonist (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75580 detects IL-1 Receptor Antagonist (human) in Not Applicable samples. RP-75580 has been successfully used in Control and Western Blot procedures. RP-75580 The immunogen is N/A.