Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IL-1 beta (human) Recombinant, Thermo Scientific™

IL-1 beta (Hu) Recombinant protein RP-75586 from Thermo Scientific™ specifically detects IL-1 beta (Hu) in Not Applicable samples, and it is validated for Control and WB

$272.60 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight ∽21.88
Name Human IL-1 beta Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75584

 
pierce biotechnology inc
RP75584
10μg purified protein (0.05mg/mL) in Laemmli and no preservative 10μg Each for $272.60

PIRP75585

 
pierce biotechnology inc
RP75585
100μg purified protein (1mg/mL) in TBS with 50% glycerol and no preservative; pH 8 100μg Each for $759.80

PIRP75586

 
pierce biotechnology inc
RP75586
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight ∽21.88
Name Human IL-1 beta Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements Store at -20°C

IL-1 beta (Hu) Recombinant protein RP-75586 from Thermo Scientific™ specifically detects IL-1 beta (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75586 detects IL-1 beta (human) in Not Applicable samples. RP-75586 has been successfully used in Control and Western Blot procedures. RP-75586 The immunogen is N/A.