Please login to your online account to display your discounted pricing

anti-IL-1 beta (human) Recombinant, Thermo Scientific™


IL-1 beta (Hu) Recombinant protein RP-75586 from Thermo Scientific™ specifically detects IL-1 beta (Hu) in Not Applicable samples, and it is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75586

 View more versions of this product

 Disclaimers

in vitro research only

Catalog No. PIRP75586

  • / Each
 

Description & Specifications

Specifications

Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Host Species E. Coli
Conjugate Unlabeled
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Immunogen N/A
pH Range 8
Name Human IL-1 beta Recombinant Protein
Molecular Weight ∽21.88
Source E. coli
Cross Reactivity Human
Recombinant Yes
Quantity 1mg

IL-1 beta (Hu) Recombinant protein RP-75586 from Thermo Scientific™ specifically detects IL-1 beta (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Formulation: 1mg purified protein in PBS with 50% glycerol and no preservative
RP-75586 contains purified recombinant human IL-1 beta protein with an N-terminal hexahistidine tag. RP-75586 may be used as a positive control in many immunological assays.