Please login to your online account to display your discounted pricing

IGF1 Recombinant Protein, Invitrogen™


IGF1 Recombinant Protein, RP-10910, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP10910

Catalog No. PIRP10910

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control
Formulation 100μg purified protein in 0.05M acetic acid and no preservative
Host Species E. Coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Gene Alias IGF-I, IGF-1, Somatomedin C
Format Purified
Purity >98%
Name IGF-1 Recombinant Protein
Purification Method SDS-PAGE
Recombinant Yes
Source E. coli
Quantity 100μg

IGF1 Recombinant Protein, RP-10910, from Invitrogen.


Host Species: E. Coli
Formulation: 100μg purified protein in 0.05M acetic acid and no preservative
The RP-10910 protein is a recombinant protein suitable for use as a control in various applications. Purity: Greater than 98% by RP-HPLC, anion-exchange chromatography; reducing and non-reducing SDS-PAGE, silver stain. Dimers/Aggregates: <1% by SDSPAGE Biological Activity: Fully biologically active compared to reference standards. Specific activity is 1 x 106 IU/mg. Biological activity at 0.2-20 ng/ml. Endotoxin: <0.1 ng/ug Protein Content: Determined spectrophotometrically at 280nm (0.502ext. coeff), and by RP-HPLC analysis against a calibrated standard. Lyophilized product is stable at room temperature for 3 weeks. Store lyophilized product desiccated at -20°C. Upon reconstitution, store at 4°C for 2-7 days or aliquot and store at -20°C. For long-term storage add either HSA or BSA to 0.1%. Avoid repeated freeze-thaw cycles.