Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IGF1 Recombinant, Thermo Scientific™

IGF1 Recombinant protein RP-10910 from Thermo Scientific™ specifically detects IGF1 in Not Applicable samples, and it is validated for Control

$622.05 - $622.05

Specifications

Format Purified
For Use With (Application) Control
Name IGF-1 Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitroresearch only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP10931

 
pierce biotechnology inc
RP10931
1000μg purified protein in 0.05M acetic acid and no preservative 1000μg Each for $622.05
Description & Specifications

Specifications

Format Purified
For Use With (Application) Control
Name IGF-1 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements Store at -20°C

IGF1 Recombinant protein RP-10910 from Thermo Scientific™ specifically detects IGF1 in Not Applicable samples, and it is validated for Control


Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 100μg purified Protein in 0.05M acetic acid, with no preservative;
Synonyms: IGF-I, IGF-1, Somatomedin C

RP-10910 detects IGF1 in Not Applicable samples. RP-10910 has been successfully used in Control procedures. RP-10910