Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-CD54/ICAM-1 Recombinant, Thermo Scientific™

CD54/ICAM-1 Recombinant protein RP-75643 from Thermo Scientific™ specifically detects CD54/ICAM-1 in Not Applicable samples, and it is validated for Control and Western Blot

$870.00 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control
Name Human CD54/ICAM-1 Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75642

 
pierce biotechnology inc
RP75642
100μg purified protein (0.16mg/mL) in PBS with 50% glycerol and no preservative 100μg Each for $870.00

PIRP75643

 
pierce biotechnology inc
RP75643
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control
Name Human CD54/ICAM-1 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

CD54/ICAM-1 Recombinant protein RP-75643 from Thermo Scientific™ specifically detects CD54/ICAM-1 in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: BB2; P3.58; ICAM-1; cell surface glycoprotein P3.58; intercellular adhesion molecule 1; human rhinovirus receptor; major group rhinovirus receptor

RP-75643 detects CD54/ICAM-1 in Not Applicable samples. RP-75643 has been successfully used in Control and Western Blot procedures. RP-75643 The immunogen is N/A.