Please login to your online account to display your discounted pricing

HIV1 Reverse Transcriptase Recombinant Protein, Invitrogen™


HIV1 Reverse Transcriptase Recombinant Protein, RP-4913, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP4913

Catalog No. ARC4913

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Format Purified
Formulation 1000 units purified protein (0.37mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol, 5mMmgCl2, 0.1% Triton X-100 and no preservative; pH 8.3
Molecular Weight ∽66 and ∽51
Name HIV1 Reverse Transcriptase Recombinant Protein
pH Range 8.3
Purification Method Chromatography
Quantity 1000 UN
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Conjugate Unlabeled
Host Species E. coli
Recombinant Yes

HIV1 Reverse Transcriptase Recombinant Protein, RP-4913, from Invitrogen.

HIV-1 reverse transcriptase is an RNA-dependent DNA polymerase of HIV-1 (AIDS virus), subtype B origin. It also has RNaseH activity and is an enzyme indispensable to the reproduction of AIDS virus.
Host Species: E. Coli
Formulation: 1000 units purified protein (0.37mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol, 5mMmgCl2, 0.1% Triton X-100 and no preservative; pH 8.3
RP-4913 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 Reverse Transcriptase (PA1-4948, PA1-4949, or PA1-4950), as well as in screening of specific inhibitors for new AIDS treatment . RP-4913 is composed of two subunits with calculated molecular weights of ∽66 kDa and ∽51 kDa. RP-4913 has a specific activity of 10,000-20,000 units/mg . Purity is 90%.