Please login to your online account to display your discounted pricing

HIV1 Reverse Transcriptase Recombinant Protein, Invitrogen™


HIV1 Reverse Transcriptase Recombinant Protein, RP-4912, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP4912

Catalog No. ARC4912

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Quantity 200 UN
Formulation 200 units purified protein (4000 units/ml) in TBS with 50% glycerol, 0.01M beta mercaptoethanol, 5mMmgCl2, 0.1% Triton X-100 and no preservative; pH 8.3
Host Species E. Coli
Conjugate Unlabeled
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name HIV1 Reverse Transcriptase Recombinant Protein
Recombinant Yes
Purification Method Chromatography
Concentration 4000U/mL
Molecular Weight ∽66 and ∽51
pH Range 8.3
Source E. coli

HIV1 Reverse Transcriptase Recombinant Protein, RP-4912, from Invitrogen.

HIV-1 reverse transcriptase is an RNA-dependent DNA polymerase of HIV-1 (AIDS virus), subtype B origin. It also has RNaseH activity and is an enzyme indispensable to the reproduction of AIDS virus.
Host Species: E. Coli
Formulation: 200 units purified protein (4000 units/ml) in TBS with 50% glycerol, 0.01M beta mercaptoethanol, 5mMmgCl2, 0.1% Triton X-100 and no preservative; pH 8.3
RP-4912 can be used as a positive control in Western blotting with ABR antibodies againt -HIV-1 Reverse Transcriptase, as well as in screening of specific inhibitors for new AIDS treatment . RP-4912 is composed of two subunits with calculated molecular weights of ∽66 kDa and ∽51 kDa; see figure 1. RP-4912 has a specific activity of ∽4,000 units/ml