Please login to your online account to display your discounted pricing

HIV1 p55 Recombinant Protein, Invitrogen™


HIV1 p55 Recombinant Protein, RP-4921, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP4921

Catalog No. ARC4921

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, ELISA, Western Blot
Formulation 100μg purified protein (0.44mg/mL) in TBS with 20% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
Quantity 100μg
Host Species E. Coli
Conjugate Unlabeled
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Molecular Weight ∽55
Purification Method Chromatography
Name HIV1 p55 Recombinant Protein
pH Range 7.5
Concentration 0.5mg/mL
Source E. coli
Recombinant Yes

HIV1 p55 Recombinant Protein, RP-4921, from Invitrogen.

HIV-1 Gag p55 is a precursor protein of several proteins that form the core structure of AIDS virus, indispensable to its reproduction. This protein is digested by HIV-1 protease, first into intermediate products p41 and p15. Then p41 is digested into matrix protein p17 and capsid protein p24. Protein p15 is further digested into nucleocapsid protein p7 and to p6 and p1, both of unknown function.
Host Species: E. Coli
Formulation: 100μg purified protein (0.44mg/mL) in TBS with 20% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
RP-4921 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 p55 (PA1-4959, or PA1-4960), as well as a p55 antigen in detection of anti-HIV-1 p55 antibody in Western blotting or ELISA. Also, it can be used as standard for the quantitative analysis of HIV-1 p55 antigen. RP-4921 has a calculated molecular weight of ∽55 kDa. Purity is 90%.