Please login to your online account to display your discounted pricing

HIV1 p24 Recombinant Protein, Invitrogen™


HIV1 p24 Recombinant Protein, RP-4916, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP4916

Catalog No. ARC4916

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, ELISA, Western Blot
Formulation 20μg purified protein (1mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
Quantity 20μg
Host Species E. Coli
Conjugate Unlabeled
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Recombinant Yes
Molecular Weight ∽24
Name HIV1 p24 Recombinant Protein
Purification Method Chromatography
pH Range 7.5
Concentration 0.48mg/mL
Source E. coli

HIV1 p24 Recombinant Protein, RP-4916, from Invitrogen.

HIV-1 Gag p24 is a capsid protein that constitutes the core of AIDS virus HIV-1 and is processed by the digestion of its precursor Gag p55 by HIV-1 protease. This protein is indispensable to the reproduction of AIDS virus and constitutes an essential element in the virus particle construction. As this protein is detectable from the early stage of AIDS virus infection, it is used as a marker for observation of the development in the patient’s condition after treatment, as it indicates the amount of virus in the blood.
Host Species: E. Coli
Formulation: 20μg purified protein (1mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
RP-4916 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 p24 (PA1-4951, PA1-4952, or PA1-4953), as well as a p24 antigen in detection of anti-HIV-1 p24 antibody in Western blotting or ELISA. Also, it can be used as standard in the titration of p24 antigens as it indicates the AIDS virus amount in virus infection, diagnosis and treatment progress RP-4916 has a calculated molecular weight of ∽24 kDa. Purity is 90%.