Please login to your online account to display your discounted pricing

HIV1 p17 Recombinant Protein, Invitrogen™


HIV1 p17 Recombinant Protein, RP-4915, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP4915

Catalog No. ARC4915

  • / Each
 

Description & Specifications

Specifications

Concentration 0.48mg/mL
For Use With (Application) Control, ELISA, Western Blot
Format Purified
Formulation 100μg purified protein in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
Molecular Weight ∽17
Name HIV1 p17 Recombinant Protein
pH Range 7.5
Purification Method Chromatography
Quantity 100μg
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Conjugate Unlabeled
Host Species E. coli
Recombinant Yes

HIV1 p17 Recombinant Protein, RP-4915, from Invitrogen.

HIV-1 Gag p17 is the matrix protein of AIDS virus HIV-1 and is processed by the digestion of its precursor Gag p55 by HIV-1 protease. This protein is indispensable to the reproduction of AIDS virus and constitute the essential element of the of AIDS virus particle construction.
Host Species: E. Coli
Formulation: 100μg purified protein in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
RP-4915 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 p17 (PA1-4954, PA1-4955, or PA1-4956), as well as a p17 antigen in detection of anti-HIV-1 p17 antibody in Western blotting or ELISA. Also, it can be used as standard for the quantitative analysis of HIV-1 p17 antigen. RP-4915 has a calculated molecular weight of ∽17 kDa. Purity is 90%.