Please login to your online account to display your discounted pricing

HIV1 p15 Recombinant Protein, Invitrogen™


HIV1 p15 Recombinant Protein, RP-4918, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP4918

Catalog No. ARC4918

  • / Each
 

Description & Specifications

Specifications

Quantity 20μg
Conjugate Unlabeled
For Use With (Application) Control, ELISA, Western Blot
Formulation 20μg purified protein (0.42mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
Host Species E. Coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Molecular Weight ∽15
Name HIV1 p15 Recombinant Protein
pH Range 7.5
Purification Method Chromatography
Recombinant Yes
Source E. coli
Concentration 0.48mg/mL

HIV1 p15 Recombinant Protein, RP-4918, from Invitrogen.

HIV-1 Gag p15 is processed by the digestion of its precursor Gag p55 by HIV-1 protease. This protein is further digested into nucleocapsid protein p7 and into p6 and p1 of unknown function. This digestion is promoted by the binding of HIV-1 genome RNA and the two Zn finger motifs that exist in the p7 region. The produced nucleocapsid protein p7 regulates the RNA function by directly binding to HIV-1 genome RNA.
Host Species: E. Coli
Formulation: 20μg purified protein (0.42mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
RP-4918 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 p15 (PA1-4957, or PA1-4958), as well as a p15 antigen in detection of anti-HIV-1 p15 antibody in Western blotting or ELISA. Also, it can be used as standard for the quantitative analysis of HIV-1 p15 antigen. RP-4918 has a calculated molecular weight of ∽15 kDa. Purity is 90%.