Please login to your online account to display your discounted pricing

HIV1 nef Recombinant Protein, Invitrogen™


HIV1 nef Recombinant Protein, RP-4923, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP4923

Catalog No. ARC4923

  • / Each
 

Description & Specifications

Specifications

Quantity 100μg
Conjugate Unlabeled
For Use With (Application) Control, ELISA, Western Blot
Formulation 100μg purified protein (0.48mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
Host Species E. Coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Molecular Weight ∽27
Name HIV1 nef Recombinant Protein
pH Range 7.5
Purification Method Chromatography
Recombinant Yes
Source E. coli
Concentration 0.48mg/mL

HIV1 nef Recombinant Protein, RP-4923, from Invitrogen.

HIV-1 Nef is one of the accessory proteins synthesized in the early stage of AIDS virus reproduction and is abundantly found in infected cells. The name derives from its negative factor thought at the beginning but presently it is remarked as the protein which bears a most distinctive biological characteristic of AIDS virus. The protein interacts directly with the signal transduction protein of the host T cell and works effectively on AIDS infection or on long term survival of the infected cells or induces apoptosis of non-infected cells. It is also involved in the endocytosis and degradation of receptor protein of the cell surface such as CD4 and MH4, important for AIDS virus infection.
Host Species: E. Coli
Formulation: 100μg purified protein (0.48mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
RP-4923 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 Nef (PA1-4961, PA1-4962, or PA1-4963), as well as a HIV-1Nef antigen in detection of anti-HIV-1 Nef antibody in Western blotting or ELISA. RP-4923 has a calculated molecular weight of ∽27 kDa. Purity is 90%.