Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Haptoglobin b-chain (human) Recombinant, Thermo Scientific™

Haptoglobin b-chain (Hu) Recombinant protein RP-75577 from Thermo Scientific™ detects Haptoglobin b-chain (Hu) in Not Applicable samples, and is validated for Control and WB

$361.05 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Haptoglobin beta chain Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75575

 
pierce biotechnology inc
RP75575
10μg purified protein in Laemmli and no preservative; pH 6.8 10μg Each for $361.05

PIRP75576

 
pierce biotechnology inc
RP75576
100μg purified protein in TBS with 50% glycerol and no preservative 100μg Each for $800.40

PIRP75577

 
pierce biotechnology inc
RP75577
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Haptoglobin beta chain Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Haptoglobin b-chain (Hu) Recombinant protein RP-75577 from Thermo Scientific™ detects Haptoglobin b-chain (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75577 detects Haptoglobin b-chain (human) in Not Applicable samples. RP-75577 has been successfully used in Control and Western Blot procedures. RP-75577 The immunogen is N/A.