Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-GST Omega 1 140D (human) Recombinant, Thermo Scientific™

GST Omega 1 140D (Hu) Recombinant protein RP-75568 from Thermo Scientific™ detects GST Omega 1 140D (Hu) in Not Applicable samples, and is validated for Control and WB

$272.60 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human GST omega 1 140D Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75566

 
pierce biotechnology inc
RP75566
10μg purified protein in 0.02M tris with 50% glycerol and no preservative; pH 8.0 10μg Each for $272.60

PIRP75568

 
pierce biotechnology inc
RP75568
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75567

 
pierce biotechnology inc
RP75567
100μg purified protein in 0.02M tris with 50% glycerol and no preservative; pH 8.0 100μg Each for $685.85
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human GST omega 1 140D Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

GST Omega 1 140D (Hu) Recombinant protein RP-75568 from Thermo Scientific™ detects GST Omega 1 140D (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75568 detects GST Omega 1 140D (human) in Not Applicable samples. RP-75568 has been successfully used in Control and Western Blot procedures. RP-75568 The immunogen is N/A.