Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-GST Omega 1 140A (human) Recombinant, Thermo Scientific™

GST Omega 1 140A (Hu) Recombinant protein RP-75565 from Thermo Scientific™ detects GST Omega 1 140A (Hu) in Not Applicable samples, and is validated for Control and WB

$272.60 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human GST omega 1 140A Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75563

 
pierce biotechnology inc
RP75563
10μg purified protein in 0.02M tris with 50% glycerol and no preservative; pH 8 10μg Each for $272.60

PIRP75564

 
pierce biotechnology inc
RP75564
100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $685.85

PIRP75565

 
pierce biotechnology inc
RP75565
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human GST omega 1 140A Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

GST Omega 1 140A (Hu) Recombinant protein RP-75565 from Thermo Scientific™ detects GST Omega 1 140A (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75565 detects GST Omega 1 140A (human) in Not Applicable samples. RP-75565 has been successfully used in Control and Western Blot procedures. RP-75565 The immunogen is N/A.