Please login to your online account to display your discounted pricing

anti-Granulocyte Macrophage-CSF (human) Recombinant, Thermo Scientific™


Granulocyte Macrophage-CSF (Hu) Recombinant protein RP-75574 detects Granulocyte Macrophage-CSF (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75572

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75572

  • / Each
 

Description & Specifications

Specifications

Formulation 10μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
For Use With (Application) Control, Western Blot
Conjugate Unlabeled
Host Species E. Coli
Format Purified
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Immunogen N/A
Gene Alias Granulocyte Macrophage Colony Stimulating Factor
pH Range 8
Name Human Granulocyte Macrophage-Colony Stimulating Factor (GM)-CSF Recombinant Protein
Recombinant Yes
Cross Reactivity Human
Quantity 10μg
Source E. coli

Granulocyte Macrophage-CSF (Hu) Recombinant protein RP-75574 detects Granulocyte Macrophage-CSF (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 10μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
RP-75572 contains purified recombinant human GM-Colony Stimulating Factor protein with an N-terminal hexahistidine tag. RP-75572 may be used as a positive control in many immunological assays.