Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Granulocyte Macrophage-CSF (human) Recombinant, Thermo Scientific™

Granulocyte Macrophage-CSF (Hu) Recombinant protein RP-75574 detects Granulocyte Macrophage-CSF (Hu) in Not Applicable samples, and is validated for Control and WB

$272.60 - $3,915.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Granulocyte Macrophage-Colony Stimulating Factor (GM)-CSF Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75572

 
pierce biotechnology inc
RP75572
10μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8 10μg Each for $272.60

PIRP75574

 
pierce biotechnology inc
RP75574
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,915.00

PIRP75573

 
pierce biotechnology inc
RP75573
100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8 100μg Each for $759.80
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Granulocyte Macrophage-Colony Stimulating Factor (GM)-CSF Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Granulocyte Macrophage-CSF (Hu) Recombinant protein RP-75574 detects Granulocyte Macrophage-CSF (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Granulocyte Macrophage Colony Stimulating Factor

RP-75574 detects Granulocyte Macrophage-CSF (human) in Not Applicable samples. RP-75574 has been successfully used in Control and Western Blot procedures. RP-75574 The immunogen is N/A.