Please login to your online account to display your discounted pricing

Granulocyte Macrophage-CSF (human) Recombinant Protein, Invitrogen™


Granulocyte Macrophage-CSF (human) Recombinant Protein, RP-75574, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP75574

Catalog No. PIRP75574

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Format Purified
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
Name Human Granulocyte Macrophage-Colony Stimulating Factor (GM)-CSF Recombinant Protein
Quantity 1mg
Source E. coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Gene Alias Granulocyte Macrophage Colony Stimulating Factor
Conjugate Unlabeled
Cross Reactivity Human
Host Species E. coli
Recombinant Yes

Granulocyte Macrophage-CSF (human) Recombinant Protein, RP-75574, from Invitrogen.


Host Species: E. Coli
Formulation: 1mg purified protein in PBS with 50% glycerol and no preservative
RP-75574 contains purified recombinant human GM-Colony Stimulating Factor protein with an N-terminal hexahistidine tag. RP-75574 may be used as a positive control in many immunological assays.