Please login to your online account to display your discounted pricing

anti-Granulocyte Macrophage-CSF (human) Recombinant, Thermo Scientific™


Granulocyte Macrophage-CSF (Hu) Recombinant protein RP-75574 detects Granulocyte Macrophage-CSF (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75573

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75573

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Gene Alias Granulocyte Macrophage Colony Stimulating Factor
Format Purified
Name Human Granulocyte Macrophage-Colony Stimulating Factor (GM)-CSF Recombinant Protein
pH Range 8
Quantity 100μg
Recombinant Yes
Source E. coli
Cross Reactivity Human

Granulocyte Macrophage-CSF (Hu) Recombinant protein RP-75574 detects Granulocyte Macrophage-CSF (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
RP-75573 contains purified recombinant human GM-Colony Stimulating Factor protein with an N-terminal hexahistidine tag. RP-75573 may be used as a positive control in many immunological assays.