Please login to your online account to display your discounted pricing

anti-GCSF (human) Recombinant, Thermo Scientific™


GCSF (human) Recombinant protein RP-75571 from Thermo Scientific™ specifically detects GCSF (human) in Not Applicable samples, and it is validated for Control and Western Blot

Manufacturer: pierce biotechnology inc  RP75569

 View more versions of this product

 Disclaimers

in vitro research purpose only.

Catalog No. PIRP75569

  • / Each
 

Description & Specifications

Specifications

Formulation 10μg purified protein in tris HCl with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Conjugate Unlabeled
Host Species E. Coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Gene Alias Granulocyte Colony Stimulating Factor
Immunogen N/A
Recombinant Yes
Name Human GCSF Recombinant Protein
Molecular Weight 23.19
Quantity 10μg
Cross Reactivity Human
Source E. coli

GCSF (human) Recombinant protein RP-75571 from Thermo Scientific™ specifically detects GCSF (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Formulation: 10μg purified protein in tris HCl with 50% glycerol and no preservative
RP-75569 contains purified recombinant human GCSF protein with an N-terminal hexahistidine tag. RP-75569 may be used as a positive control in many immunological assays.