Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-GCSF (human) Recombinant, Thermo Scientific™

GCSF (human) Recombinant protein RP-75571 from Thermo Scientific™ specifically detects GCSF (human) in Not Applicable samples, and it is validated for Control and Western Blot

$272.60 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 23.19
Name Human GCSF Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitro research purpose only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75569

 
pierce biotechnology inc
RP75569
10μg purified protein in tris HCl with 50% glycerol and no preservative 10μg Each for $272.60

PIRP75570

 
pierce biotechnology inc
RP75570
100μg purified protein (0.8mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0 100μg Each for $797.50

PIRP75571

 
pierce biotechnology inc
RP75571
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 23.19
Name Human GCSF Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

GCSF (human) Recombinant protein RP-75571 from Thermo Scientific™ specifically detects GCSF (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Granulocyte Colony Stimulating Factor

RP-75571 detects GCSF (human) in Not Applicable samples. RP-75571 has been successfully used in Control and Western Blot procedures. RP-75571 The immunogen is N/A.