Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Fibroblast Growth Factor basic (human) Recombinant, Thermo Scientific™

Fibroblast Growth Factor basic (Hu) Recombinant protein RP-8627 detects Fibroblast Growth Factor basic (Hu) in Not Applicable samples, and is validated for Control

$384.25 - $390.05

Specifications

Concentration 0.1mg/mL
Cross Reactivity Human
Format Purified
Formulation 50μg purified protein (0.1mg/mL) in 10mM sodium phosphate and no preservative; pH 8.0
For Use With (Application) Control
View More Specs
Products
Catalog Number Mfr. No. Molecular Weight Price Quantity    

PIRP8628

 
pierce biotechnology inc
RP8628
∽17.2 Each for $390.05

PIRP8627

 
pierce biotechnology inc
RP8627
∽16.5 Each for $384.25
Description & Specifications

Specifications

Concentration 0.1mg/mL
Cross Reactivity Human
Format Purified
Formulation 50μg purified protein (0.1mg/mL) in 10mM sodium phosphate and no preservative; pH 8.0
For Use With (Application) Control
Name Human Fibroblast Growth Factor basic Recombinant Protein
pH Range 8
Quantity 50μg
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Fibroblast Growth Factor basic (Hu) Recombinant protein RP-8627 detects Fibroblast Growth Factor basic (Hu) in Not Applicable samples, and is validated for Control

Fibroblast Growth Factors (FGFs) are a 22 member family of proteins known to be involved in angiogenesis, wound healing and embryonic development. As a family, they bind to heparin and signal through four receptor tyrosine kinases called, FGFR1, 2, 3 and 4. Although the mechanism remains unclear, FGF-basic 147 (variant of FGF basic 154), also called FGF-2, is a critical component in keeping embryonic stem cells undifferentiated in cell culture systems.
Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 50μg purified Protein (0.1mg/mL) in 10mM sodium phosphate, with no preservative; 8.0
Synonyms: H-FGF-b-147
Uniprot Accession: P09038
Gene Symbol: FGF2
Entrez GeneID: 2247
RP-8627 detects Fibroblast Growth Factor basic (human) in Not Applicable samples. RP-8627 has been successfully used in Control procedures. RP-8627