Please login to your online account to display your discounted pricing

anti-Epidermal Growth Factor Recombinant, Thermo Scientific™


Epidermal Growth Factor Recombinant protein RP-10927 from Thermo Scientific™ specifically detects Epidermal Growth Factor in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  RP10914

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP10914

  • / Each
 

Description & Specifications

Specifications

Formulation 500μg purified protein and no preservative
For Use With (Application) Control
Conjugate Unlabeled
Host Species E. Coli
Storage Requirements Store at -20°C
Immunogen N/A
Gene Alias Epidermal Growth Factor
Format Purified
Name Epidermal Growth Factor Recombinant Protein
Recombinant Yes
Quantity 500μg
Source E. coli

Epidermal Growth Factor Recombinant protein RP-10927 from Thermo Scientific™ specifically detects Epidermal Growth Factor in Not Applicable samples, and is validated for Control


Host Species: E. Coli
Formulation: 500μg purified protein and no preservative
The RP-10914 protein is a recombinant protein suitable for use as a control in ELISA and Western blot applications. RP-10914 is reported to have the additional biological activity: ED50 as determined by the dose-dependent proliferation of mouse 3T3 cells <0.1ng/ml. Store lyophilized product at -20C. Upon reconstitution, store at 4C for 1 week or aliquot and store at -20C.