Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Epidermal Growth Factor Recombinant, Thermo Scientific™

Epidermal Growth Factor Recombinant protein RP-10927 from Thermo Scientific™ specifically detects Epidermal Growth Factor in Not Applicable samples, and is validated for Control

$436.45 - $436.45

Specifications

Format Purified
For Use With (Application) Control
Name Epidermal Growth Factor Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP10914

 
pierce biotechnology inc
RP10914
500μg purified protein and no preservative 500μg Each for $436.45
Description & Specifications

Specifications

Format Purified
For Use With (Application) Control
Name Epidermal Growth Factor Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements Store at -20°C

Epidermal Growth Factor Recombinant protein RP-10927 from Thermo Scientific™ specifically detects Epidermal Growth Factor in Not Applicable samples, and is validated for Control


Host Species: E. Coli
Cross Reactivity: Not Applicable
Clone: N/A
Immunogen: N/A
Formulation: 1000μg purified Protein (1mg/mL) no preservative;
Synonyms: Epidermal Growth Factor

RP-10927 detects Epidermal Growth Factor in Not Applicable samples. RP-10927 has been successfully used in Control procedures. RP-10927 The immunogen is N/A.