Please login to your online account to display your discounted pricing

anti-Epidermal FABP (human) Recombinant, Thermo Scientific™


Epidermal FABP (Hu) Recombinant protein RP-75554 from Thermo Scientific™ specifically detects Epidermal FABP (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75554

 View more versions of this product

 Disclaimers

in vitroresearch only

Catalog No. PIRP75554

  • / Each
 

Description & Specifications

Specifications

Formulation 10μg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Host Species E. Coli
Conjugate Unlabeled
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Gene Alias Fatty Acid Binding Protein
Immunogen N/A
Source E. coli
Name Human Epidermal FABP Recombinant Protein
Recombinant Yes
Cross Reactivity Human
Quantity 10μg

Epidermal FABP (Hu) Recombinant protein RP-75554 from Thermo Scientific™ specifically detects Epidermal FABP (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 10μg purified protein in PBS with 50% glycerol and no preservative
RP-75554 contains purified recombinant human epidermal FABP protein with an N-terminal hexahistidine tag. RP-75554 may be used as a positive control in many immunological assays.