Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Epidermal FABP (human) Recombinant, Thermo Scientific™

Epidermal FABP (Hu) Recombinant protein RP-75554 from Thermo Scientific™ specifically detects Epidermal FABP (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $471.25

Specifications

Cross Reactivity Human
Formulation 10μg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Epidermal FABP Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitroresearch only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75554

 
pierce biotechnology inc
RP75554
10μg Each for $288.55

PIRP75555

 
pierce biotechnology inc
RP75555
100μg Each for $471.25
Description & Specifications

Specifications

Cross Reactivity Human
Formulation 10μg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Epidermal FABP Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Epidermal FABP (Hu) Recombinant protein RP-75554 from Thermo Scientific™ specifically detects Epidermal FABP (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 10μg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Fatty Acid Binding Protein

RP-75554 detects Epidermal FABP (human) in Not Applicable samples. RP-75554 has been successfully used in Control and Western Blot procedures. RP-75554 The immunogen is N/A.