Please login to your online account to display your discounted pricing

anti-Eotaxin (human) Recombinant, Thermo Scientific™


Eotaxin (Hu) Recombinant protein RP-75538 from Thermo Scientific™ specifically detects Eotaxin (Hu) in Not Applicable samples, and it is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75537

 View more versions of this product

 Disclaimers

in vitroresearch only

Catalog No. PIRP75537

  • / Each
 

Description & Specifications

Specifications

Quantity 100μg
Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 100μg purified protein in tris with 50% glycerol and no preservative; pH 8
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human Eotaxin Recombinant Protein
Source E. coli
Recombinant Yes
Cross Reactivity Human

Eotaxin (Hu) Recombinant protein RP-75538 from Thermo Scientific™ specifically detects Eotaxin (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Formulation: 100μg purified protein in tris with 50% glycerol and no preservative; pH 8
RP-75537 contains purified recombinant human eotaxin protein with an N-terminal hexahistidine tag. RP-75537 may be used as a positive control in many immunological assays.