Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Eotaxin (human) Recombinant, Thermo Scientific™

Eotaxin (Hu) Recombinant protein RP-75538 from Thermo Scientific™ specifically detects Eotaxin (Hu) in Not Applicable samples, and it is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Eotaxin Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitroresearch only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75537

 
pierce biotechnology inc
RP75537
100μg Each for $688.75

PIRP75536

 
pierce biotechnology inc
RP75536
10μg Each for $288.55

PIRP75538

 
pierce biotechnology inc
RP75538
1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Eotaxin Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Eotaxin (Hu) Recombinant protein RP-75538 from Thermo Scientific™ specifically detects Eotaxin (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75538 detects Eotaxin (human) in Not Applicable samples. RP-75538 has been successfully used in Control and Western Blot procedures. RP-75538 The immunogen is N/A.