Please login to your online account to display your discounted pricing

anti-E-Selectin (human) Recombinant, Thermo Scientific™


E-Selectin (Hu) Recombinant protein RP-75541 from Thermo Scientific™ specifically detects E-Selectin (Hu) in Not Applicable samples, and it is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75540

 View more versions of this product

Catalog No. PIRP75540

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 100μg purified protein (0.14mg/mL) in PBS with 50% glycerol and no preservative
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human E-Selectin Recombinant Protein
Quantity 100μg
Recombinant Yes
Source E. coli
Cross Reactivity Human

E-Selectin (Hu) Recombinant protein RP-75541 from Thermo Scientific™ specifically detects E-Selectin (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Formulation: 100μg purified protein (0.14mg/mL) in PBS with 50% glycerol and no preservative
RP-75540 contains purified recombinant human e-Selectin protein with an N-terminal hexahistidine tag. RP-75540 may be used as a positive control in many immunological assays.