Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-E-Selectin (human) Recombinant, Thermo Scientific™

E-Selectin (Hu) Recombinant protein RP-75541 from Thermo Scientific™ specifically detects E-Selectin (Hu) in Not Applicable samples, and it is validated for Control and WB

$272.60 - $3,770.00

Specifications

Cross Reactivity Human
Format Purified
Formulation 100μg purified protein (0.14mg/mL) in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human E-Selectin Recombinant Protein
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75539

 
pierce biotechnology inc
RP75539
10μg Each for $272.60

PIRP75540

 
pierce biotechnology inc
RP75540
100μg Each for $870.00

PIRP75541

 
pierce biotechnology inc
RP75541
1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
Format Purified
Formulation 100μg purified protein (0.14mg/mL) in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human E-Selectin Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

E-Selectin (Hu) Recombinant protein RP-75541 from Thermo Scientific™ specifically detects E-Selectin (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75541 detects E-Selectin (human) in Not Applicable samples. RP-75541 has been successfully used in Control and Western Blot procedures. RP-75541 The immunogen is N/A.