Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Cytokeratin 18 (human) Recombinant, Thermo Scientific™

CytokeRtin 18 (Hu) Recombinant protein RP-75535 from Thermo Scientific™ specifically detects CytokeRtin 18 (Hu) in Not Applicable samples, and it is validated for Control and WB

$361.05 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Cytokeratin 18 Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitroresearch only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75533

 
pierce biotechnology inc
RP75533
10μg purified protein (0.22mg/mL) in 0.02M tris HCl with 5mM EDTA, 50% glycerol and no preservative; pH 8.0 10μg Each for $361.05

PIRP75534

 
pierce biotechnology inc
RP75534
100μg purified protein (0.22mg/mL) in tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8 100μg Each for $785.90

PIRP75535

 
pierce biotechnology inc
RP75535
1mg purified protein in 20mM tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Cytokeratin 18 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

CytokeRtin 18 (Hu) Recombinant protein RP-75535 from Thermo Scientific™ specifically detects CytokeRtin 18 (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Ck18

RP-75535 detects Cytokeratin 18 (human) in Not Applicable samples. RP-75535 has been successfully used in Control and Western Blot procedures. RP-75535 The immunogen is N/A.