Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Cdk4 (human) Recombinant, Thermo Scientific™

Cdk4 (human) Recombinant protein RP-75532 from Thermo Scientific™ specifically detects Cdk4 (human) in Not Applicable samples, and it is validated for Control and Western Blot

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human CDK4 Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitroresearch only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75530

 
pierce biotechnology inc
RP75530
10μg purified protein in 10mM Bis-tris propane with 50% glycerol, 100mM NaCl and no preservative; pH 9.0 10μg Each for $288.55

PIRP75531

 
pierce biotechnology inc
RP75531
100μg purified protein (0.081mg/mL) in 0.01M Bis-tris propane with 0.1M NaCl, 50% glycerol and no preservative; pH 9.0 100μg Each for $870.00

PIRP75532

 
pierce biotechnology inc
RP75532
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human CDK4 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Cdk4 (human) Recombinant protein RP-75532 from Thermo Scientific™ specifically detects Cdk4 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Cyclin-Dependant Kinase 4

RP-75532 detects Cdk4 (human) in Not Applicable samples. RP-75532 has been successfully used in Control and Western Blot procedures. RP-75532 The immunogen is N/A.