Please login to your online account to display your discounted pricing

anti-CCL2 (human) Recombinant, Thermo Scientific™


CCL2 (human) Recombinant protein RP-75664 from Thermo Scientific™ specifically detects CCL2 (human) in Not Applicable samples, and it is validated for Control and Western Blot

Manufacturer: pierce biotechnology inc  RP75663

 View more versions of this product

 Disclaimers

in vitroresearch only

Catalog No. PIRP75663

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Formulation 100μg purified protein in PBS with 50% glycerol and no preservative
Conjugate Unlabeled
Host Species E. Coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Gene Alias Monocyte Chemotactic Protein-1, MCP1
Immunogen N/A
Name Human CCL2 Recombinant Protein
Source E. coli
Recombinant Yes
Cross Reactivity Human
Quantity 100μg

CCL2 (human) Recombinant protein RP-75664 from Thermo Scientific™ specifically detects CCL2 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Formulation: 100μg purified protein in PBS with 50% glycerol and no preservative
RP-75663 contains purified recombinant human CCL2 protein with an N-terminal hexahistidine tag. RP-75663 may be used as a positive control in many immunological assays.