Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-CCL2 (human) Recombinant, Thermo Scientific™

CCL2 (human) Recombinant protein RP-75664 from Thermo Scientific™ specifically detects CCL2 (human) in Not Applicable samples, and it is validated for Control and Western Blot

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
Name Human CCL2 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
View More Specs

 Disclaimers

in vitroresearch only

Products
Catalog Number Mfr. No. For Use With (Application) Formulation Quantity Price Quantity    

PIRP75662

 
pierce biotechnology inc
RP75662
Control, Western Blot 10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75663

 
pierce biotechnology inc
RP75663
Control, Western Blot 100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $797.50

PIRP75664

 
pierce biotechnology inc
RP75664
Control, Western Blot 1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP8648

 
pierce biotechnology inc
RP8648
Control 20μg purified protein (0.1mg/mL) in water and no preservative 20μg Each for $448.05
Description & Specifications

Specifications

Cross Reactivity Human
Name Human CCL2 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

CCL2 (human) Recombinant protein RP-75664 from Thermo Scientific™ specifically detects CCL2 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Monocyte Chemotactic Protein-1, MCP1

RP-75664 detects CCL2 (human) in Not Applicable samples. RP-75664 has been successfully used in Control and Western Blot procedures. RP-75664 The immunogen is N/A.