Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-CAIII (human) Recombinant, Thermo Scientific™

CAIII (human) Recombinant protein RP-75526 from Thermo Scientific™ specifically detects CAIII (human) in Not Applicable samples, and it is validated for Control and Western Blot

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human CAIII Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitroresearch only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75524

 
pierce biotechnology inc
RP75524
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75525

 
pierce biotechnology inc
RP75525
100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $723.55

PIRP75526

 
pierce biotechnology inc
RP75526
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human CAIII Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

CAIII (human) Recombinant protein RP-75526 from Thermo Scientific™ specifically detects CAIII (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Carbonic Anhydrase III

RP-75526 detects CAIII (human) in Not Applicable samples. RP-75526 has been successfully used in Control and Western Blot procedures. RP-75526 The immunogen is N/A.