Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-C-Reactive Protein (human) Recombinant, Thermo Scientific™

C-Reactive Protein (Hu) Recombinant protein RP-75529 from Thermo Scientific™ detects C-Reactive Protein (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $797.50

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human C-Reactive Protein Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitroresearch only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75529

 
pierce biotechnology inc
RP75529
1mg purified protein in TBS with 2mM CaCl2 and 0.05% sodium azide 1mg Each for $797.50

PIRP75527

 
pierce biotechnology inc
RP75527
10μg purified protein in 20mM tris HCl with 0.14M NaCl, 2mM CaCl2 and 0.05% sodium azide; pH 7.5 10μg Each for $288.55

PIRP75528

 
pierce biotechnology inc
RP75528
100μg purified protein (1mg/mL) in TBS with 2mM CaCl2 and 0.05% sodium azide; pH 7.5 100μg Each for $303.05
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human C-Reactive Protein Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements Store at 4° C

C-Reactive Protein (Hu) Recombinant protein RP-75529 from Thermo Scientific™ detects C-Reactive Protein (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in TBS, with 2mM CaCl2 and 0.05% sodium azide;

RP-75529 detects C-Reactive Protein (human) in Not Applicable samples. RP-75529 has been successfully used in Control and Western Blot procedures. RP-75529 The immunogen is N/A.