Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Brain Fatty Acid Binding Protein (human) Recombinant, Thermo Scientific™

Brain Fatty Acid Binding Protein (Hu) Recombinant protein RP-75542 detects Brain Fatty Acid Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $448.05

Specifications

Cross Reactivity Human
Formulation 10μg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Brain Fatty Acid Binding Protein Recombinant Protein
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75543

 
pierce biotechnology inc
RP75543
100μg Each for $448.05

PIRP75542

 
pierce biotechnology inc
RP75542
10μg Each for $288.55
Description & Specifications

Specifications

Cross Reactivity Human
Formulation 10μg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Brain Fatty Acid Binding Protein Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Brain Fatty Acid Binding Protein (Hu) Recombinant protein RP-75542 detects Brain Fatty Acid Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 10μg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Fatty Acid Binding Protein

RP-75542 detects Brain Fatty Acid Binding Protein (human) in Not Applicable samples. RP-75542 has been successfully used in Control and Western Blot procedures. RP-75542 The immunogen is N/A.