Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Goat Gamma Globulin Control, Thermo Scientific™

Gt Gamma Globulin Control isotype control 31871 from Thermo Scientific™ detects Gt Gamma Globulin in Not Applicable samples, and is validated for CHIP, Control, FACS and IP

$67.20 - $144.90

Products
Catalog Number Mfr. No. For Use With (Application) Cross Reactivity Host Species Product Type Price Quantity    

PI31887

 
pierce biotechnology inc
31887
Each for $67.20

PI31871

 
pierce biotechnology inc
31871
Each for $81.90

PI31878

 
pierce biotechnology inc
31878
Pack for $144.90

PI31879

 
pierce biotechnology inc
31879
Each for $81.90

PI31885

 
pierce biotechnology inc
31885
Each for $98.96
Description & Specifications

Gt Gamma Globulin Control isotype control 31871 from Thermo Scientific™ detects Gt Gamma Globulin in Not Applicable samples, and is validated for CHIP, Control, FACS and IP


Host Species: Goat
Cross Reactivity: Not Applicable
Isotype: Ig Mix
Formulation: 10 mg purified Isotype Control (11.7mg/mL) in PBS, with no preservative; 7.6
Synonyms: Negative Control, Isotype Control, Immunoglobulin Control, Control

MA5-16458 detects NKG2D/KLRK1 in Human samples. MA5-16458 has been successfully used in Flow Cytometry procedures. MA5-16458 The immunogen is NKL cells..