Please login to your online account to display your discounted pricing
Thermo Scientific™ Owl™ EasyCast™ Mini Gel Electrophoresis System Promotion
Buy select lab essential equipment for $150 each
$760.94 - $760.94
Specifications
Certifications/Compliance CE marked
Product Type Mini
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    
09-528-110
  Accessories
thermo scientific™
B1A
Each for $760.94
Description & Specifications
Specifications
Certifications/Compliance CE marked
Product Type Mini
The EasyCast systems feature leakproof casting that allows a gel to be cast and run in the same chamber with no tape or additional parts required. The EasyCast B2's longer gel length and the 2 comb slots provide the flexibility of running 8 to 48 samples on 1 gel.
  • Two comb slots on gel tray double sample capacity by doubling number of sample wells
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands