Please login to your online account to display your discounted pricing
Thermo Scientific™ Orion™ Test Kits for AQUAfast™ IV Colorimeters
$148.40 - $148.40
Products
Catalog Number Mfr. No. Price Quantity    
13-642-164
thermo scientific™
ORIAC4006
Each for $148.40
Description & Specifications
  • Kits offer premeasured reagents in bar-coded Auto-Test™ cuvets
  • AQUAfast IV Colorimeter has an Auto-ID™ function that reads the bar code to identify the species of interest automatically, then selects proper method, wavelength and time
  • Just read results on built-in display
  • Cuvet is opened only when measurement is taken, so reagent quality is not affected