Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Streamline classroom preparation by allocating buffer pouches to each work station.
$88.45 - $89.53
Chemical Identifiers
Products 2
Catalog Number Mfr. No. Quantity pH Price Quantity  
Catalog Number Mfr. No. Quantity pH Price Quantity  
S04959 View Documents Thermo Scientific™
910410WAEMD
10 Pouches 4.01 Pack of 10 Each for $89.53
S04972 View Documents Thermo Scientific™
911010EMD
10 Pouches 10.00 Pack of 10 Each for $88.45
Description
Description
Eliminate spills and ensure easy cleanup using calibration buffers in the pouch.
  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST™ traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs
  • Buffers are also available in pint (475mL) bottles, portable five packs of 2 oz. (5 x 60mL) bottles, convenient All-in-One buffer kits and 5 gallon (19L) cubitainers
Specifications
Chemical Identifiers
0.0
icon-pdf.png
Specifications
4.01
10 Pouches