Learn More
Thermo Scientific™ Orion™ ROSS™ Sure-Flow™ pH Electrode with Sure-flow junction, BNC connector, PROMO
Measure general pH samples as well as viscous or colloidal samples with the Thermo Scientific™ Orion™ 8172BNWP ROSS™ Sure-Flow™ pH Electrode.
Manufacturer: Thermo Scientific™ 8172BNWPPM
Description
Measure general pH samples as well as viscous or colloidal samples with the free-flowing sleeve junction design of the Thermo Scientific™ Orion™ 8172BNWP ROSS™ Sure-Flow™ pH Electrode.
This general purpose pH electrode offers superior performance for dirty, difficult samples such as soils, sludges, colloids, viscous materials and organics. Junction cleans easily between measurements; pressing down on the cap allows filling solution to flow through the junction carries away any contamination that may interfere with measurement.
- Material: Glass body
- pH range: 0 to 14
- Temperature range: 0° to 100°C
- Internal reference: ROSS
- Junction: Sure-Flow
- Dimensions: 120 × 12 mm
- pH precision: 0.01
- Fill solution: 3M KCl, Orion Cat. No 810007
- Cap diameter: 16 mm
Includes:
- Detachable bulb guard, two bottles of Orion ROSS electrode fill solution
Specifications
Orion ROSS Sure-Flow pH electrode | |
3M KCl (810007) | |
Detachable bulb guard, 2 x 60mL bottles of ORion ROSS electrode fill solution | |
0.01 | |
BNC Waterproof | |
Glass Body | |
Glass Body |
16 mm | |
Biological/Pharmaceutical ?TRIS buffer, proteins, enzymes; Emulsions ? Foods, cosmetics, oils; Emulsions ? Petroleum products, paint; General Purpose ? Most sample types; High Ionic Strength ? Acids, bases, brines, pH > 12 or pH < 2; Low Ionic Strength ? | |
ROSS | |
32°F to 212°F | |
Sure-Flow | |
0°C to 100°C | |
0 to 14 |
Promotions
-
Save 30% on Thermo Scientific Industrial Lab Items
Get 30% off select products for sample preparation and analysis in industrial laboratories.: 06/30/21