Please login to your online account to display your discounted pricing
Thermo Scientific™ Nicolet™ iS™5 FT-IR Spectrometer
No-compromise FT-IR Performance, Fit and Affordability
Products
Catalog Number Mfr. No. Price Quantity    
14-200-121
thermo scientific™
912A0712
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
The Thermo Scientific Nicolet iS5 FT-IR spectrometer provides superior FT-IR performance in a compact size at an affordable price. Combining flexible sample handling and leading Thermo Scientific OMNIC™ software, the Nicolet iS5 spectrometer is the ideal FT-IR solution for industrial, government and academic labs around the world.
The Thermo Scientific Nicolet iS5 FT-IR spectrometer provides the ideal performance and fit for product assurance testing and material identification. It implements the same field-proven FT-IR technology found on other Thermo Scientific Nicolet FT-IR models and is driven by award-winning Thermo Scientific OMNIC software.
The lightweight design and small footprint of the Nicolet iS5 FT-IR spectrometer enables you to bring the spectrometer where you need answers most: from your busy laboratory to the production floor or in the warehouse. Additionally, the spectrometer's form factor and low cost-of-ownership make it a perfect match for analytical laboratories seeking to deploy multiple units across manufacturing facilities worldwide.
Premier performance in a compact size
The Nicolet iS5 spectrometer features a footprint similar in size to a laptop computer, making it small enough and light enough to move around easily.
  • 10kg (22 lb.) weight makes it extremely transportable
  • Rugged enough to transport around the plant or across campus
  • Footprint: 35x 28cm (13.5 x 10.9 inches)
Durable design
The Nicolet iS5 spectrometer is designed to provide trouble-free operation even in challenging environments.
  • Field-proven optics encased in a sturdy magnesium-alloy frame
  • A thermally-controlled diode laser provides many years of worry-free use
  • User-accessible parts minimize service costs
Easy to maintain
Replacing the IR source or desiccant in your spectrometer does not require a service call, or even opening up the instrument.
Ready to handle any environment
  • Withstands a wide range of temperature and humidity conditions
  • Vibration, electromagnetic interference (EMI), dust, and even tilt were all considered in the design of this instrument Magnesium-alloy construction provides excellent mechanical characteristics for stiffness, thermal properties, vibrational dampening and reduced weight