Thermo Scientific™ National™ Headspace 20mm Crimp Seals with Septa
National Crimp Seals ensure sample security every time.
Manufacturer:  Thermo Scientific™ C402032APK
Catalog No. 03-377-297
Description
Description
20mm Crimp seals with prefitted septa for headspace vials
  • Seals with pre-inserted septa are convenient and minimize contamination due to handling
  • Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
  • PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
  • Molded polyethylene SepCap™ is suitable for short-term storage or where repeated access to the vial is required
SepCap is for short-term storage. Do not use SepCap at elevated temperatures.
Order Info
Septa and crimp seals also sold separately.
Specifications
Specifications
Silica Glass
Septa
Septa
Amber
Documents