Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ StarHead™ Magnetic Stir Bars
Greater mixing action at bottom, side, and top of container, even at low stirring speeds
$73.32 - $73.32
Products
Catalog Number Mfr. No. Price Quantity    
14-511-98A
thermo scientific™
NDS66000010
Case for $73.32
Description & Specifications
For test tubes, round-bottom or flat-bottom labware.
  • TFE construction with star-shaped fins and encapsulated Alnico-V magnets
  • Chemical and high-temperature (to 260°C [500°F]) resistant
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.